Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Diethylether 99+% stabilised, pure

Glass bottle, 1 L
  ACRO123990010
 
 
Diethylether 99+% stabilised, pure
  ACRO123990010
 :  60-29-7
Diethylether 99+% stabilised, pure
Diethylether 99+% stabilised, pure

Glass bottle, 1 L

 :  60-29-7
 
494,00 kr
   
1 * 1 L
 
 :  Ether
  •  :60-29-7
  •  :(CH₃CH₂)₂O
  •  :MFCD00011646
  •  :74.12 g/mol
  •  :12,03852
  •  :34,6 °C (760 mmHg)
  •  :–116 °C
  •  :0,714 g/cm³
  •  :–45 °C
  •  :1155
  •  :3,I
  •  :stabiliseret med BHT/Ionol
  •  :Stue temperatur
  •  :200-467-2
  •  :01-2119535785-29
 

 

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3510 to 1.3540 (20°C, 589 nm)
  • Water:
    ≤0.2 % (Coulometric)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    5 to 10 ppm BHT
  • Residue after evaporation:
    ≤0.002 %
  • Peroxides:
    ≤0.001 %

 

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.