Iodacetamid 98%
Plastic bottle, 100 g
: 2-Iodoacetamide
- :144-48-9
- :ICH₂CONH₂
- :MFCD00008028
- :184.96 g/mol
- :266 °C (1013 hPa)
- :>90 °C
- :Køleskab
- :205-630-1
- :2923
- :8,II
- Appearance:White to yellow to beige Powder or crystals
- Infrared spectrum:Conforms
- Melting point:≥90 °C
- Titration Argentometric:≥97.5 % (ex I)
- Pakning:Plastic bottle
- Pakn:100 g



