Iodacetamid 98%
Glass bottle, 25 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: 2-Iodoacetamide
- CAS nummer:144-48-9
- Formel:ICH₂CONH₂
- MDL nummer:MFCD00008028
- Molecular Weight:184.96 g/mol
- Kogende Pt:266 °C (1013 hPa)
- Smeltende Pt:>90 °C
- Opbevaringstemperatur:Køleskab
- EINECS:205-630-1
- FN:2923
- ADR:8,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to yellow to beige Powder or crystals
- Infrared spectrum:Conforms
- Melting point:≥90 °C
- Titration Argentometric:≥97.5 % (ex I)
Specifikationer
- Pakning:Glass bottle
- Pakn:25 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



