8-Quinolinol ACS
Glass bottle for solids, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: 8-Hydroxyquinoline
- CAS nummer:148-24-3
- Formel:C₉H₇NO
- MDL nummer:MFCD00006807
- Molecular Weight:145.16 g/mol
- Merck Index:13,04869
- Kogende Pt:267 °C (752 mmHg)
- Smeltende Pt:72…74 °C
- Tæthed:1,03 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:205-711-1
- FN:2811
- ADR:6.1,III
Documents
ACRO12190
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White to light yellow to light beige Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:72.5 to 74.0 °C
- Residue after ignition:≤0.05 %
- Suitability:(for Mg determination) Passes test
- Sulfate (SO4):≤0.02 %
- Insoluble matter:≤0.05 % (in alcohol)
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g



