Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

trans-4-Hydroxy-L(-)-prolin 99+%

Glass bottle for solids, 100 g
  ACRO121781000
trans-4-Hydroxy-L(-)-prolin 99+%
  ACRO121781000
 :  51-35-4
trans-4-Hydroxy-L(-)-prolin 99+%
trans-4-Hydroxy-L(-)-prolin 99+%

Glass bottle for solids, 100 g

 :  51-35-4
 
2.710,00 kr
   
1 * 100 g
 
 :  L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
  •  :51-35-4
  •  :C₅H₉NO₃
  •  :MFCD00064320
  •  :131.13 g/mol
  •  :13,04866
  •  :296 °C (1013 hPa)
  •  :274…275 °C (dec.)
  •  :Stue temperatur
  •  :200-091-9
 

 

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.2 % (105°C, 3 hrs)
  • Sulfated ash:
    ≤0.1 %
  • UV transmittance:
    ≥95 % (c=10, H2O, 430 nm) 1 cm cell
  • Specific optical rotation:
    -74° to -77° (20°C, 589 nm) (c=5, H2O)
  • Titration with HClO4:
    99.0 to 101.0 %
  • Chloride (Cl):
    ≤200 ppm
  • Sulfate (SO4):
    ≤200 ppm
  • Ammonium (NH4):
    ≤200 ppm
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Other amino acids:
    ≤0.5 % (TLC)

 

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.