Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
trans-4-Hydroxy-L(-)-prolin 99+%

trans-4-Hydroxy-L(-)-prolin 99+%

Glass bottle for solids, 25 g
Katalog # ACRO121780250
Leverandør:   Thermo Fisher Scientific
trans-4-Hydroxy-L(-)-prolin 99+%
Katalog # ACRO121780250
CAS nummer:  51-35-4
trans-4-Hydroxy-L(-)-prolin 99+%
trans-4-Hydroxy-L(-)-prolin 99+%

Glass bottle for solids, 25 g

CAS nummer:  51-35-4
Price
956,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
  • CAS nummer:51-35-4
  • Formel:C₅H₉NO₃
  • MDL nummer:MFCD00064320
  • Molecular Weight:131.13 g/mol
  • Merck Index:13,04866
  • Kogende Pt:296 °C (1013 hPa)
  • Smeltende Pt:274…275 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-091-9

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.2 % (105°C, 3 hrs)
  • Sulfated ash:
    ≤0.1 %
  • UV transmittance:
    ≥95 % (c=10, H2O, 430 nm) 1 cm cell
  • Specific optical rotation:
    -74° to -77° (20°C, 589 nm) (c=5, H2O)
  • Titration with HClO4:
    99.0 to 101.0 %
  • Chloride (Cl):
    ≤200 ppm
  • Sulfate (SO4):
    ≤200 ppm
  • Ammonium (NH4):
    ≤200 ppm
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Other amino acids:
    ≤0.5 % (TLC)

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.