Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

n-Hexadecan 99%, pure

Glass bottle, 500 mL
Katalog # ACRO120465000
Sundhed
Leverandør:   Thermo Fisher Scientific
n-Hexadecan 99%, pure
Katalog # ACRO120465000
CAS nummer:  544-76-3
n-Hexadecan 99%, pure
n-Hexadecan 99%, pure

Glass bottle, 500 mL

CAS nummer:  544-76-3
Price
1.470,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Synonymer:  Hexadecane, Cetane
  • CAS nummer:544-76-3
  • Formel:H₃C(CH₂)₁₄CH₃
  • MDL nummer:MFCD00008998
  • Molecular Weight:226.45 g/mol
  • Kogende Pt:287 °C (760 mmHg)
  • Smeltende Pt:18 °C
  • Tæthed:0,773 g/cm³
  • Flash Pt:135 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:208-878-9
Specifikationer

Specifikationstestresultater

  • Appearance:
    Clear colorless Liquid or solid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4330 to 1.4350 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL

Specifikationer