n-Hexadecan 99%, pure
Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
n-Hexadecan 99%, pure
Catalog # ACRO120461000
Supplier: Thermo Fisher Scientific
CAS Number: 544-76-3
Jump to:
Product Details & Documents
Synonyms: Hexadecane, Cetane
- CAS Number:544-76-3
- Formula:H₃C(CH₂)₁₄CH₃
- MDL Number:MFCD00008998
- Molecular Weight:226.45 g/mol
- Boiling Pt:287 °C (760 mmHg)
- Melting Pt:18 °C
- Density:0,773 g/cm³
- Flash Pt:135 °C
- Storage Temperature:Stue temperatur
- EINECS:208-878-9
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid or solid
- Infrared spectrum:Conforms
- Refractive index:1.4330 to 1.4350 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pakning:Glass bottle
- Pakn:100 mL



