EDTA 99%, pure
Plastic bottle, 250 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
- CAS nummer:60-00-4
- Formel:C₁₀H₁₆N₂O₈
- MDL nummer:MFCD00003541
- Molecular Weight:292.25 g/mol
- Merck Index:13,03546
- Kogende Pt:614 °C (1013 hPa)
- Smeltende Pt:220 °C (dec.)
- Tæthed:0,86 g/cm³ (20 °C)
- Flash Pt:>100 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-449-4
- NÅ:01-2119486399-18
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:237 to 245 °C
- Titration Complexometric:≥98.5 %
- pH:(sat. soln. in water) Approx. 2 to 3
- Nitrate (NO3):≤0.5 ppm
- Ammonium (NH4):≤2 ppm
- Arsenic (As):≤0.01 ppm
- Iron (Fe):≤0.2 ppm
- Mercury (Hg):≤5 ppb
- Heavy metals (as Pb):≤1 ppm
- KMnO4-reducing substance:≤2 ppm (as SO2)
Specifikationer
- Pakning:Plastic bottle
- Pakn:250 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



