Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Estrone 99+%

Estrone 99+%

Glass bottle, 5 g
Katalog # ACRO117840050
Sundhed
Leverandør:   Thermo Fisher Scientific
Estrone 99+%
Katalog # ACRO117840050
CAS nummer:  53-16-7
Estrone 99+%
Estrone 99+%

Glass bottle, 5 g

CAS nummer:  53-16-7
Price
1.850,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
  • CAS nummer:53-16-7
  • Formel:C₁₈H₂₂O₂
  • MDL nummer:MFCD00003620
  • Smeltende Pt:258…261 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-164-5

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White to almost white Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    258 to 261 °C
  • Loss on drying:
    ≤0.5 % (105 °C, 3 hrs)
  • Sulfated ash:
    ≤0.5 %
  • Additional info:
    limit of equilenin and equilin: passes test
  • HPLC:
    ≥99.0 % (on dried substance)
  • Specific optical rotation:
    +158° to +165° (20 °C, 589 nm) (c=1, dioxane)
  • Clarity of solution:
    passes test
  • Silicon dioxide (SiO2):
    ≤0.005 %
  • Sulfur compounds (as SO4):
    ≤0.004 %
  • Insoluble matter:
    ≤0.01 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    5 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.