(±)-Epichlorhydrin 99%
Glass bottle, 25 mL
Leverandør: Thermo Fisher Scientific
(±)-Epichlorhydrin 99%
Katalog # ACRO117780250
Leverandør: Thermo Fisher Scientific
CAS nummer: 106-89-8
Gå til:
Product Details & Documents
Synonymer: Epichlorhydrin, 1-Chlor-2,3-epoxypropan, (±)-1-Chlor-2,3-epoxypropan, 3-Chlor-1,2-epoxypropan, (±)-3-Chlor-1,2-epoxypropan
- CAS nummer:106-89-8
- Formel:C₃H₅ClO
- MDL nummer:MFCD00005132
- Molecular Weight:92.52 g/mol
- Merck Index:13,03642
- Kogende Pt:115…117 °C (760 mmHg)
- Smeltende Pt:–57 °C
- Tæthed:1,18 g/cm³
- Flash Pt:28 °C
- Opbevaringstemperatur:Køleskab
- EINECS:203-439-8
- FN:2023
- ADR:6.1,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4365 to 1.4385 (20°C, 589 nm)
- Water:≤0.02 % (K.F.)
- GC:≥98.5 %
- Color scale:≤15 APHA
- Silicate and phosphate:≤5 ppm (as SiO2)
- Iron (Fe):≤1 ppm
- Heavy metals (as Pb):≤1 ppm
- Nitrogen compounds (as N):≤0.001 %
Specifikationer
- Pakning:Glass bottle
- Pakn:25 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



