Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dipropylamin 99%

Dipropylamin 99%

Glass bottle, 250 mL
Katalog # ACRO117492500
Brandfarlig
Ætsende
Giftig
Leverandør:   Thermo Fisher Scientific
Dipropylamin 99%
Katalog # ACRO117492500
CAS nummer:  142-84-7
Dipropylamin 99%
Dipropylamin 99%

Glass bottle, 250 mL

CAS nummer:  142-84-7
Price
191,00 kr
Log ind for tilgængelighed
1 * 250 mL
Product Details & Documents
Synonymer:  N-Propyl-1-propanamine
  • CAS nummer:142-84-7
  • Formel:C₆H₁₅N
  • MDL nummer:MFCD00009362
  • Molecular Weight:101.19 g/mol
  • Merck Index:12,03406
  • Kogende Pt:109,4 °C (1013 hPa)
  • Smeltende Pt:–63 °C
  • Tæthed:0,74 g/cm³
  • Flash Pt:7 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:205-565-9
  • FN:2383
  • ADR:3,II

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4030 to 1.4050 (20°C, 589 nm)
  • Water:
    ≤0.2 %
  • GC:
    ≥98.5 %
  • Color scale:
    ≤15 APHA
  • Heavy metals (ICP-OES):
    ≤5 ppm
  • Insoluble matter:
    ≤0.005 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    250 mL

Specifikationer