Dipropylamin 99%
Glass bottle, 1 L
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: N-Propyl-1-propanamine
- CAS nummer:142-84-7
- Formel:C₆H₁₅N
- MDL nummer:MFCD00009362
- Molecular Weight:101.19 g/mol
- Merck Index:12,03406
- Kogende Pt:109,4 °C (1013 hPa)
- Smeltende Pt:–63 °C
- Tæthed:0,74 g/cm³
- Flash Pt:7 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:205-565-9
- FN:2383
- ADR:3,II
Documents
ACRO11749
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4030 to 1.4050 (20°C, 589 nm)
- Water:≤0.2 %
- GC:≥98.5 %
- Color scale:≤15 APHA
- Heavy metals (ICP-OES):≤5 ppm
- Insoluble matter:≤0.005 %
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
- Cat. No.:ACRO117490010
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



