Cyclopentanon 99+%, pure
Glass bottle, 1 L
Leverandør: Thermo Fisher Scientific
Cyclopentanon 99+%, pure
Katalog # ACRO111530010
Leverandør: Thermo Fisher Scientific
CAS nummer: 120-92-3
Gå til:
Product Details & Documents
- CAS nummer:120-92-3
- Formel:C₅H₈O
- MDL nummer:MFCD00001409
- Molecular Weight:84.12 g/mol
- Merck Index:13,02771
- Kogende Pt:130…131 °C (760 mmHg)
- Smeltende Pt:–51 °C
- Tæthed:0,95 g/cm³
- Flash Pt:26 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-435-9
- FN:2245
- ADR:3,III
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4360 to 1.4380 (20°C, 589 nm)
- GC:≥99.0 %
- Stabilizer:0.01 % alpha-Tocopherol
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L



