3-Hydroxybenzonitril 97%
Glass bottle for solids, 50 g
Leverandør: Thermo Fisher Scientific
3-Hydroxybenzonitril 97%
Katalog # ACRO110820500
Leverandør: Thermo Fisher Scientific
CAS nummer: 873-62-1
Gå til:
Product Details & Documents
Synonymer: m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol
- CAS nummer:873-62-1
- Formel:C₇H₅NO
- MDL nummer:MFCD00002252
- Molecular Weight:119.12 g/mol
- Kogende Pt:265 °C (1013 hPa)
- Smeltende Pt:75…80 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:212-845-4
- FN:3439
- ADR:6.1,III
Documents
ACRO11082
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:75 to 80 °C
- GC:≥96.0 %
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:50 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



