3-Hydroxybenzonitril 97%
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
3-Hydroxybenzonitril 97%
Catalog # ACRO110820100
Supplier: Thermo Fisher Scientific
CAS Number: 873-62-1
Jump to:
Product Details & Documents
Synonyms: m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol
- CAS Number:873-62-1
- Formula:C₇H₅NO
- MDL Number:MFCD00002252
- Molecular Weight:119.12 g/mol
- Boiling Pt:265 °C (1013 hPa)
- Melting Pt:75…80 °C
- Storage Temperature:Stue temperatur
- EINECS:212-845-4
- UN:3439
- ADR:6.1,III
Documents
ACRO11082
SDS
Specifications
Specification Test Results
- Appearance:White to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:75 to 80 °C
- GC:≥96.0 %
Specifications
- Pakning:Glass bottle for solids
- Pakn:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



