Cumol 98%
Glass bottle, 1 L
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Cumen
- CAS nummer:98-82-8
- Formel:C₉H₁₂
- MDL nummer:MFCD00008881
- Molecular Weight:120.19 g/mol
- Merck Index:13,02644
- Kogende Pt:151 °C (1013 hPa)
- Smeltende Pt:–96 °C
- Tæthed:0,862 g/cm³
- Flash Pt:31 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-704-5
- FN:1918
- ADR:3,III
Documents
ACRO11063
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4890 to 1.4930 (20°C, 589 nm)
- GC:≥97.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



