m-Cresol 99%
Glass bottle for solids, 1 kg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: m-Methylphenol
- CAS nummer:108-39-4
- Formel:CH₃C₆H₄OH
- MDL nummer:MFCD00002302
- Molecular Weight:108.14 g/mol
- Merck Index:13,02604
- Kogende Pt:203 °C (760 mmHg)
- Smeltende Pt:8…10 °C
- Tæthed:1,03 g/cm³
- Flash Pt:86 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-577-9
- FN:2076
- ADR:6.1,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light red Viscous liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.5390 to 1.5420 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg



