Coronen 95%
Glass bottle for solids, 100 mg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Hexabenzobenzene
- CAS nummer:191-07-1
- Formel:C₂₄H₁₂
- MDL nummer:MFCD00004134
- Molecular Weight:300.36 g/mol
- Kogende Pt:525 °C (1013 hPa)
- Smeltende Pt:438…440 °C
- Tæthed:1,371 g/cm³ (25 °C)
- EINECS:205-881-7
- FN:2811
Documents
ACRO11051
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Yellow to green Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
- Titration RCOCl:≥96.0 %
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 mg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



