Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Congo red indicator grade

Congo red indicator grade

Glass bottle, 500 g
Katalog # ACRO110505000
Sundhed
Leverandør:   Thermo Fisher Scientific
Congo red indicator grade
Katalog # ACRO110505000
CAS nummer:  573-58-0
Congo red indicator grade
Congo red indicator grade

Glass bottle, 500 g

CAS nummer:  573-58-0
Price
2.990,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  CI 22120, Dinatrium-3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalen-1-sulfonat)
  • CAS nummer:573-58-0
  • Formel:C₃₂H₂₂N₆Na₂O₆S₂
  • MDL nummer:MFCD00004028
  • Molecular Weight:696.67 g/mol
  • Merck Index:12,02562
  • Smeltende Pt:>360 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:209-358-4

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Dark red to red-brown Powder
  • Infrared spectrum:
    Conforms
  • UV spectrum:
    Complies
  • Solubility:
    (0.1 % in water) Clear
  • Visual transition interval:
    from pH 3.0 (violet) to pH 5.2 (red-orange)

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 g

Specifikationer