(2-Hydroxyethyl)trimethylammoniumchlorid 99%
Plastic bottle, 500 g
Leverandør: Thermo Fisher Scientific
(2-Hydroxyethyl)trimethylammoniumchlorid 99%
Katalog # ACRO110295000
Leverandør: Thermo Fisher Scientific
CAS nummer: 67-48-1
Gå til:
Product Details & Documents
Synonymer: 2-hydroxy-N,N,N-trimethylethanaminium chloride
- CAS nummer:67-48-1
- Formel:HOH₂CCH₂N(CH₃)₃Cl
- MDL nummer:MFCD00011721
- Molecular Weight:139.62 g/mol
- Merck Index:13,02226
- Smeltende Pt:298…304 °C (dec.)
- Tæthed:~ 1,205 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-655-4
Specifikationer
Specifikationstestresultater
- Appearance:White Adhering crystals
- Infrared spectrum:Conforms
- Titration Argentometric:98.5 to 101.5 % (on dry substance)
- Water:≤2 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



