Glycin, Millipore®
Glass bottle for solids, 500 g
Katalog # 3570-500
Leverandør: Merck Millipore (Calbiochem)
Gå til:
Product Details & Documents
Synonymer: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS nummer:56-40-6
- Formel:NH₂CH₂COOH
- MDL nummer:MFCD00008131
- Molecular Weight:75.07 g/mol
- Merck Index:13,04504
- Kogende Pt:181 °C (1013 hPa)
- Smeltende Pt:83 °C
- Tæthed:1,64 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-272-2
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



