Glycin, Millipore®
Glass bottle for solids, 1 kg
Katalog # 3570-1
Leverandør: Merck Millipore (Calbiochem)
Gå til:
Product Details & Documents
Synonymer: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS nummer:56-40-6
- Formel:NH₂CH₂COOH
- MDL nummer:MFCD00008131
- Molecular Weight:75.07 g/mol
- Merck Index:13,04504
- Kogende Pt:181 °C (1013 hPa)
- Smeltende Pt:83 °C
- Tæthed:1,64 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-272-2
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg



