Genistein, Millipore®
Glass bottle for solids, 20 mg
Katalog # 345834-20
Leverandør: Merck Millipore (Calbiochem)
Genistein, Millipore®
Katalog # 345834-20
Leverandør: Merck Millipore (Calbiochem)
CAS nummer: 446-72-0
Gå til:
Product Details & Documents
Synonymer: 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone
- CAS nummer:446-72-0
- Formel:C₁₅H₁₀O₅
- MDL nummer:MFCD00016952
- Molecular Weight:270.24 g/mol
- Smeltende Pt:297…298 °C
- EINECS:207-174-9
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:20 mg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



