Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
D-(+)-Cellobiose 98%

D-(+)-Cellobiose 98%

Glass bottle, 5 g
Catalog # ACRO108460050
D-(+)-Cellobiose 98%
Catalog # ACRO108460050
CAS Number:  528-50-7
D-(+)-Cellobiose 98%
D-(+)-Cellobiose 98%

Glass bottle, 5 g

CAS Number:  528-50-7
Price
375,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
  • CAS Number:528-50-7
  • Formula:C₁₂H₂₂O₁₁
  • MDL Number:MFCD00136034
  • Molecular Weight:342.30 g/mol
  • Melting Pt:239 °C (dec.)
  • Storage Temperature:Stue temperatur
  • EINECS:208-436-5

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.5 % (K.F.)
  • Solubility:
    (5 % in water) Clear colorless
  • HPLC:
    ≥97.5 % (ELSD)
  • Specific optical rotation:
    +33° to +35° (20°C, 589 nm) (c=1, water) (after 15 h)

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    5 g

Specifications