2-Bromethanol 97%
Glass bottle, 250 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
- CAS nummer:540-51-2
- Formel:BrH₂CCH₂OH
- MDL nummer:MFCD00002827
- Molecular Weight:124.97 g/mol
- Merck Index:13,03826
- Kogende Pt:149…150 °C (750 mmHg)
- Smeltende Pt:–80 °C
- Tæthed:1,76 g/cm³
- Flash Pt:>110 °C
- Opbevaringstemperatur:Køleskab
- EINECS:208-748-1
- FN:2927
- ADR:6.1,II
Documents
ACRO10692
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4890 to 1.4940 (20°C, 589 nm)
- GC:≥96.0 %
- Acid value:≤0.1 mg KOH/g
Specifikationer
- Pakning:Glass bottle
- Pakn:250 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



