2-Bromethanol 97%
Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:540-51-2
- Formula:BrH₂CCH₂OH
- MDL Number:MFCD00002827
- Molecular Weight:124.97 g/mol
- Merck Index:13,03826
- Boiling Pt:149…150 °C (750 mmHg)
- Melting Pt:–80 °C
- Density:1,76 g/cm³
- Flash Pt:>110 °C
- Storage Temperature:Køleskab
- EINECS:208-748-1
- UN:2927
- ADR:6.1,II
Documents
ACRO10692
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4890 to 1.4940 (20°C, 589 nm)
- GC:≥96.0 %
- Acid value:≤0.1 mg KOH/g
Specifications
- Pakning:Glass bottle
- Pakn:100 mL
- Cat. No.:ACRO106921000



