2-Bromethanol 97%
Glass bottle, 5 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
- CAS nummer:540-51-2
- Formel:BrH₂CCH₂OH
- MDL nummer:MFCD00002827
- Molecular Weight:124.97 g/mol
- Merck Index:13,03826
- Kogende Pt:149…150 °C (750 mmHg)
- Smeltende Pt:–80 °C
- Tæthed:1,76 g/cm³
- Flash Pt:>110 °C
- Opbevaringstemperatur:Køleskab
- EINECS:208-748-1
- FN:2927
- ADR:6.1,II
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4890 to 1.4940 (20°C, 589 nm)
- GC:≥96.0 %
- Acid value:≤0.1 mg KOH/g
Specifikationer
- Pakning:Glass bottle
- Pakn:5 mL



