Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Benzophenon 99%, pure

Benzophenon 99%, pure

Plastic bottle, 1 kg
Katalog # ACRO105560010
Health
Leverandør:   Thermo Fisher Scientific
Benzophenon 99%, pure
Katalog # ACRO105560010
CAS nummer:  119-61-9
Benzophenon 99%, pure
Benzophenon 99%, pure

Plastic bottle, 1 kg

CAS nummer:  119-61-9
Price
564,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Diphenylmethanone, Diphenyl ketone
  • CAS Number:119-61-9
  • Formula:C₁₃H₁₀O
  • MDL Number:MFCD00003076
  • Molecular Weight:182.22 g/mol
  • Merck Index:13,01099
  • Boiling Pt:305 °C (760 mmHg)
  • Melting Pt:47…49 °C
  • Density:1,085 g/cm³ (20 °C)
  • Flash Pt:143 °C
  • Storage Temperature:Stue temperatur
  • EINECS:204-337-6
  • ADR:9,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    47 to 49 °C
  • GC:
    ≥98.5 %

Specifications

  • Pakning:
    Plastic bottle
  • Pakn:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.