Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Benzaldehyd 98+%

Benzaldehyd 98+%

Glass bottle, 1 L
Katalog # ACRO105220010
Irriterende
Leverandør:   Thermo Fisher Scientific
Benzaldehyd 98+%
Katalog # ACRO105220010
CAS nummer:  100-52-7
Benzaldehyd 98+%
Benzaldehyd 98+%

Glass bottle, 1 L

CAS nummer:  100-52-7
Price
436,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
  • CAS nummer:100-52-7
  • Formel:C₆H₅CHO
  • MDL nummer:MFCD00003299
  • Molecular Weight:106.12 g/mol
  • Merck Index:13,01057
  • Kogende Pt:179 °C (1013 hPa)
  • Smeltende Pt:–26 °C
  • Tæthed:1,043 g/cm³
  • Flash Pt:64 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:202-860-4
  • FN:1990
  • ADR:9,III

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5440 to 1.5460 (20°C, 589 nm)
  • GC:
    ≥98.0 %
  • Specific gravity:
    (25°C) 1.041 to 1.046
  • Acid value:
    ≤5 mg KOH/g

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.