Digitonin, Millipore®
Glass bottle for solids, 250 mg
Katalog # 300410-250
Leverandør: Merck Millipore (Calbiochem)
Digitonin, Millipore®
Katalog # 300410-250
Leverandør: Merck Millipore (Calbiochem)
CAS nummer: 11024-24-1
Gå til:
Product Details & Documents
Synonymer: Digitonin
- CAS nummer:11024-24-1
- Formel:C₅₆H₉₂O₂₉
- MDL nummer:MFCD00077729
- Molecular Weight:1229.312 g/mol
- EINECS:234-255-6
- FN:3462
- ADR:6.1,III
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:250 mg



