Orders LinkContactUsLinkComponent
Density hydrometers
Density hydrometers
Hydrometer 7, 1,060 - 1,120 g/cm³, 0,001 g/cm³, 300 mm
Katalog # AMARH800146
Leverandør:   Amarell
Density hydrometers
Katalog # AMARH800146
Leverandør:  Amarell
Density hydrometers
Density hydrometers
Hydrometer 7, 1,060 - 1,120 g/cm³, 0,001 g/cm³, 300 mm
Price
294,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
General purpose hydrometers for density areas.
  • Paper scale and shot weighted body
  • Hydrometer accuracy ±1 scale division
  • Thermometer inside the hydrometer body
Used in laboratories and industry.
Specifikationer
  • Beskrivelse:
    Hydrometer 7
  • Længde:
    300 mm
  • Seq:
    37
  • Inddeling:
    0,001 g/cm³
  • Område:
    1,060 - 1,120 g/cm³