Orders LinkContactUsLinkComponent
Density hydrometers
Density hydrometers
Hydrometer 4, 0,880 - 0,940 g/cm³, 0,001 g/cm³, 160 mm
Catalog # AMARH800010
Supplier:   Amarell
Density hydrometers
Catalog # AMARH800010
Supplier:  Amarell
Density hydrometers
Density hydrometers
Hydrometer 4, 0,880 - 0,940 g/cm³, 0,001 g/cm³, 160 mm
Price
250,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
General purpose hydrometers for density areas.
  • Paper scale and shot weighted body
  • Hydrometer accuracy ±1 scale division
  • Thermometer inside the hydrometer body
Used in laboratories and industry.
Specifications
  • Beskrivelse:
    Hydrometer 4
  • Længde:
    160 mm
  • Seq:
    04
  • Inddeling:
    0,001 g/cm³
  • Område:
    0,880 - 0,940 g/cm³