Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Chicken Native Avidin (from Egg)

Chicken Native Avidin (from Egg)

Plastic bottle for solids, 100 mg
Catalog # 189725-100
Chicken Native Avidin (from Egg)
Catalog # 189725-100
CAS Number:  
Chicken Native Avidin (from Egg)

Plastic bottle for solids, 100 mg

CAS Number:  
Price
4.900,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents
Specifications

Specifications

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    100 mg
  • Cat. No.:
    189725-100
  • CAS-nr:
    1405-69-2
  • Protein/peptide type:
    Native
  • Protein/peptide name:
    Avidin
  • MDL No.:
    MFCD00130572
  • Kilde:
    Egg
  • Species:
    Chicken
  • Opbevaring betingelser:
    Freezer

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.