Chicken Native Avidin (from Egg)
Plastic bottle for solids, 10 mg
Katalog # 189725-10
Leverandør: Merck Millipore (Oncogene)
Chicken Native Avidin (from Egg)
Katalog # 189725-10
Leverandør: Merck Millipore (Oncogene)
CAS nummer:
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Pakning:Plastic bottle for solids
- Pakn:10 mg
- Cat. No.:189725-10
- CAS-nr:1405-69-2
- Protein/peptide type:Native
- Protein/peptide name:Avidin
- MDL No.:MFCD00130572
- Kilde:Egg
- Species:Chicken
- Opbevaring betingelser:Freezer
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



