Orders LinkContactUsLinkComponent
Standard hydrometers, M100
M100, 1,500 - 1,600 g/ml, 0,002 g/ml, 250 mm
  614-1639
Standard hydrometers, M100
  614-1639
Standard hydrometers, M100
Standard hydrometers, M100
M100, 1,500 - 1,600 g/ml, 0,002 g/ml, 250 mm
272,00 kr
1 * 1 Items
These specific gravity hydrometers are marked with reference norms, calibration temperature, surface tension, individual and serial numbers.
  • Reading below the meniscus
  • Calibration temperature of 15 °C
  • Benchmark on the stem to show any movement of the scale
Calibrated according to the following tensions:
0,600 to 1,000 Sp.Gr.= 16 to 35 mN/m (low tension)
1,000 to 1,300 Sp.Gr.= 55 mN/m (medium tension)
 : ISO 650.
 : 'EU' Traceable Calibration Certificate or 'EO' Official Calibration Certificate is available on request. Includes a Traceability and Conformity Certificate.
  • Længde:
    250 mm
  • Seq:
    09
  • Inddeling:
    0,002 g/ml
  • Model:
    M100
  • Område:
    1,500 - 1,600 g/ml