Standard hydrometers, M100
M100, 1,100 - 1,200 g/ml, 0,002 g/ml, 250 mm
Catalog # 614-1635
Supplier: Alla France
Jump to:
Product Details & Documents
These specific gravity hydrometers are marked with reference norms, calibration temperature, surface tension, individual and serial numbers.
- Reading below the meniscus
- Calibration temperature of 15 °C
- Benchmark on the stem to show any movement of the scale
Calibrated according to the following tensions:
0,600 to 1,000 Sp.Gr.= 16 to 35 mN/m (low tension)
1,000 to 1,300 Sp.Gr.= 55 mN/m (medium tension)
Certifications: ISO 650.
Ordering information: 'EU' Traceable Calibration Certificate or 'EO' Official Calibration Certificate is available on request. Includes a Traceability and Conformity Certificate.
Documents
Specifications
- Længde:250 mm
- Seq:06
- Inddeling:0,002 g/ml
- Model:M100
- Område:1,100 - 1,200 g/ml