Orders LinkContactUsLinkComponent
Calibrated Baumé hydrometers
Calibrated Baumé hydrometers
0 - 10, 0,1, 300 mm
Catalog # ALLA0101FC010/20QP
Supplier:   Alla France
Calibrated Baumé hydrometers
Catalog # ALLA0101FC010/20QP
Supplier:  Alla France
Calibrated Baumé hydrometers
Calibrated Baumé hydrometers
0 - 10, 0,1, 300 mm
Price
349,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Baumé hydrometers calibrated at 20 °C.
Specifications
  • Længde:
    300 mm
  • Seq:
    03
  • Inddeling:
    0,1
  • Område:
    0 - 10