Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bariumnitrat ≥99.95% (metals basis)

Bariumnitrat ≥99.95% (metals basis)

Glass bottle for solids, 25 g
Katalog # 30481.14
Oxiderende
Irriterende
Leverandør:   Thermo Fisher Scientific
Bariumnitrat ≥99.95% (metals basis)
Katalog # 30481.14
CAS nummer:  10022-31-8
Bariumnitrat ≥99.95% (metals basis)
Bariumnitrat ≥99.95% (metals basis)

Glass bottle for solids, 25 g

CAS nummer:  10022-31-8
Price
645,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Purity: 99.95% CAS No.: 10022-31-8 Molecular Formula: Ba(NO3)2 Formula Weight: 261.35 Physical Form: Crystalline Density: 3.24 Melting Point: 592° dec. Merck Reference: 14,983 UN No.: 1446 MDL No.: MFCD00003442

  • CAS nummer:10022-31-8
  • Formel:Ba(NO₃)₂
  • MDL nummer:MFCD00003442
  • Molecular Weight:261.34 g/mol
  • Merck Index:14,983
  • Smeltende Pt:592 °C (dec.)
  • Tæthed:3,24
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:233-020-5
  • FN:1446
  • ADR:5.1,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer