Orders LinkContactUsLinkComponent
Accessories for fluid and solid samplers
Accessories for fluid and solid samplers
Sugeslange, PE, Ø 6 mm, vægtykkelse: 1 mm
Katalog # 300-1045
Leverandør:   BURKLE
Accessories for fluid and solid samplers
Katalog # 300-1045
Leverandør:  BURKLE
Accessories for fluid and solid samplers
Accessories for fluid and solid samplers
Sugeslange, PE, Ø 6 mm, vægtykkelse: 1 mm
Price
978,00 kr
Log ind for tilgængelighed
1 * 100 m
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Sugeslange, PE, Ø 6 mm, vægtykkelse: 1 mm
  • Seq:
    10