Natriumchlorid 99+% ACS
Plastic bottle, 500 g
Leverandør: Thermo Fisher Scientific
Natriumchlorid 99+% ACS
Katalog # ACRO424295000
Leverandør: Thermo Fisher Scientific
CAS nummer: 7647-14-5
Gå til:
Product Details & Documents
Synonymer: Saline, NaCl, Sea salt, Halite
- CAS nummer:7647-14-5
- Formel:NaCl
- MDL nummer:MFCD00003477
- Molecular Weight:58.44 g/mol
- Merck Index:14,08599
- Kogende Pt:1461 °C (760 mmHg)
- Smeltende Pt:801 °C
- Tæthed:2,16 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-598-3
Specifikationer
Specifikationstestresultater
- Appearance:White Crystals
- Titration Argentometric:≥99.0 %
- Loss on drying:≤0.5 % (1 g, 105°C, 2 hrs)
- pH:5.0 to 9.0 (5 % solution at 25.0°C)
- Bromide (Br):≤0.01 %
- Iodide (I):≤0.002 %
- Chlorate and Nitrate (as NO3):≤0.003 %
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤0.004 %
- Barium (Ba):Passes test
- Calcium (Ca):≤0.002 %
- Iron (Fe):≤2 ppm
- Magnesium (Mg):≤0.001 %
- Potassium (K):≤0.005 %
- Heavy metals (ICP-OES):≤5 ppm
- Insoluble matter:≤0.005 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:500 g



