Natriumchlorid 99+% ACS
Plastic bottle, 1 kg
Leverandør: Thermo Fisher Scientific
Natriumchlorid 99+% ACS
Katalog # ACRO424290010
Leverandør: Thermo Fisher Scientific
CAS nummer: 7647-14-5
Gå til:
Product Details & Documents
Synonymer: Saline, NaCl, Sea salt, Halite
- CAS nummer:7647-14-5
- Formel:NaCl
- MDL nummer:MFCD00003477
- Molecular Weight:58.44 g/mol
- Merck Index:14,08599
- Kogende Pt:1461 °C (760 mmHg)
- Smeltende Pt:801 °C
- Tæthed:2,16 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-598-3
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White Crystals
- Titration Argentometric:≥99.0 %
- Loss on drying:≤0.5 % (1 g, 105°C, 2 hrs)
- pH:5.0 to 9.0 (5 % solution at 25.0°C)
- Bromide (Br):≤0.01 %
- Iodide (I):≤0.002 %
- Chlorate and Nitrate (as NO3):≤0.003 %
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤0.004 %
- Barium (Ba):Passes test
- Calcium (Ca):≤0.002 %
- Iron (Fe):≤2 ppm
- Magnesium (Mg):≤0.001 %
- Potassium (K):≤0.005 %
- Heavy metals (ICP-OES):≤5 ppm
- Insoluble matter:≤0.005 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



