Jod 99.8+% ACS
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:7553-56-2
- Formula:I₂
- MDL Number:MFCD00011355
- Molecular Weight:253.81 g/mol
- Merck Index:13,05036
- Boiling Pt:185 °C (760 mmHg)
- Melting Pt:113 °C
- Density:4,9 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:231-442-4
- UN:3495
- ADR:8,III
Documents
ACRO42382
SDS
Specifications
Specification Test Results
- Appearance:Grey Metallic spheres
- Titration Iodometric:≥99.8 %
- Non-volatile matter:≤0.01 %
- Chlorine and Bromine (as Cl):≤0.005 %
Specifications
- Pakning:Glass bottle for solids
- Pakn:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



