Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Jod 99.8+% ACS

Glass bottle, 5 g
Katalog # ACRO423820050
Irriterende
Sundhed
Miljø
Ætsende
Leverandør:   Thermo Fisher Scientific
Jod 99.8+% ACS
Katalog # ACRO423820050
CAS nummer:  7553-56-2
Jod 99.8+% ACS
Jod 99.8+% ACS

Glass bottle, 5 g

CAS nummer:  7553-56-2
Price
260,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
  • CAS nummer:7553-56-2
  • Formel:I₂
  • MDL nummer:MFCD00011355
  • Molecular Weight:253.81 g/mol
  • Merck Index:13,05036
  • Kogende Pt:185 °C (760 mmHg)
  • Smeltende Pt:113 °C
  • Tæthed:4,9 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-442-4
  • FN:3495
  • ADR:8,III

 

Specifikationer

Specifikationstestresultater

  • Appearance:
    Grey Metallic spheres
  • Titration Iodometric:
    ≥99.8 %
  • Non-volatile matter:
    ≤0.01 %
  • Chlorine and Bromine (as Cl):
    ≤0.005 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.