Rotary evaporating flasks
50 ml, 29/32 NS, non coated
Katalog # 531-1811
Leverandør: BUCHI
Gå til:
Product Details & Documents
Made of borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® R-210/215 rotary evaporators. Pear-shaped with NS 29.2/32 ground joint socket.
Specifikationer
- Kappestr.:29/32 NS
- Model:non coated
- Kapacitet:50 ml
- Seq:50