Ammoniumnitrat 98% ACS
Plastic bottle, 1 kg
Leverandør: Thermo Fisher Scientific
Ammoniumnitrat 98% ACS
Katalog # ACRO423350010
Leverandør: Thermo Fisher Scientific
CAS nummer: 6484-52-2
Gå til:
Product Details & Documents
- CAS nummer:6484-52-2
- Formel:NH₄NO₃
- MDL nummer:MFCD00011425
- Molecular Weight:80.04 g/mol
- Merck Index:13,00538
- Kogende Pt:230 °C (1013 hPa)
- Smeltende Pt:169 °C
- Tæthed:1,72 g/cm³
- Opbevaringstemperatur:Stue temperatur
- EINECS:229-347-8
- FN:1942
- ADR:5.1,III
Documents
ACRO42335
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Adhering crystals
- Residue after ignition:≤0.01 %
- pH:4.5 to 6.0 (5 % at 25°C)
- Titration excess NaOH:≥97.5 %
- Chloride (Cl):≤5 ppm
- Nitrite (NO2):(Limit about 5 ppm) Passes test
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤0.002 %
- Iron (Fe):≤2 ppm
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 kg



