Acetone 99.6% ACS
Glass bottle, 500 mL
Leverandør: Thermo Fisher Scientific
Acetone 99.6% ACS
Katalog # ACRO423245000
Leverandør: Thermo Fisher Scientific
CAS nummer: 67-64-1
Gå til:
Product Details & Documents
Synonymer: Propanon
- CAS nummer:67-64-1
- Formel:CH₃COCH₃
- MDL nummer:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Kogende Pt:56 °C (760 mmHg)
- Smeltende Pt:–95 °C
- Tæthed:0,79 g/cm³
- Flash Pt:–20 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-662-2
- FN:1090
- ADR:3,II
- NÅ:01-2119471330-49
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Water:≤0.5 % (K.F.)
- Solubility:(in water) Passes test
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Isopropyl alcohol:≤0.05 %
- Titrable acid:≤0.0003 meq/g
- Titrable base:≤0.0006 meq/g
- Aldehyde (HCHO):≤0.002 %
- Methanol:≤0.05 %
- KMnO4-reducing substance:Passes test
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL
- Cat. No.:ACRO423245000
- Miljøcertificering:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø



