Eddikesyre 99.7+% ACS
Glass bottle, 500 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS nummer:64-19-7
- Formel:H₃CCOOH
- MDL nummer:MFCD00036152
- Molecular Weight:60.05 g/mol
- Kogende Pt:118 °C (1013 hPa)
- Smeltende Pt:16…16,5 °C
- Tæthed:1,048 g/cm³
- Flash Pt:40 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-580-7
- FN:2789
- ADR:8,II
- NÅ:01-2119475328-30
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- GC:≥99.70 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Dilution test:Passes test
- Chloride (Cl):≤1 ppm
- Sulfate (SO4):≤1 ppm
- Iron (Fe):≤0.2 ppm
- Heavy metals (as Pb):≤0.5 ppm
- Titrable base:≤0.0004 meq/g
- KMnO4-reducing substance:Passes test
- K2Cr2O7-reducing substance:Passes test
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



