Eddikesyre 99.7+% ACS
Glass bottle, 2,5 L
: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- :64-19-7
- :H₃CCOOH
- :MFCD00036152
- :60.05 g/mol
- :118 °C (1013 hPa)
- :16…16,5 °C
- :1,048 g/cm³
- :40 °C
- :Stue temperatur
- :200-580-7
- :2789
- :8,II
- :01-2119475328-30
Documents
- Appearance:Clear liquid
- GC:≥99.70 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Dilution test:Passes test
- Chloride (Cl):≤1 ppm
- Sulfate (SO4):≤1 ppm
- Iron (Fe):≤0.2 ppm
- Heavy metals (as Pb):≤0.5 ppm
- Titrable base:≤0.0004 meq/g
- KMnO4-reducing substance:Passes test
- K2Cr2O7-reducing substance:Passes test
- Pakning:Glass bottle
- Pakn:2,5 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



