Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Thymol blå ACS

Thymol blå ACS

Glass bottle for solids, 10 g
Katalog # ACRO420930100
Leverandør:   Thermo Fisher Scientific
Thymol blå ACS
Katalog # ACRO420930100
CAS nummer:  76-61-9
Thymol blå ACS
Thymol blå ACS

Glass bottle for solids, 10 g

CAS nummer:  76-61-9
Price
kr618.00
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents
Synonymer:  Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS nummer:76-61-9
  • Formel:C₂₇H₃₀O₅S
  • MDL nummer:MFCD00005869
  • Molecular Weight:466.6 g/mol
  • Merck Index:12,09541
  • Smeltende Pt:221 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-973-3

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Dark green to dark brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Visual transition interval:
    from pH 1.2 (pink) to pH 2.8 (yellow)
  • Clarity of solution:
    passes test

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.